missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Stearate, TRC

Product Code. 30134611
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134611 500 g 500g
30011325 250 g 250g
30009452 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134611 Supplier Toronto Research Chemicals Supplier No. TRCM110345500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-04-0
Molecular Formula 2 C18 H35 O2 . Mg
Molecular Weight (g/mol) 591.24
Synonym Magnesium Stearate, Octadecanoic Acid Magnesium Salt, Stearic Acid Magnesium Salt, AFCO-Chem MGS, Aurabrite MA 76, Daiwax M, Daiwax SMO, Dibasic magnesium stearate, EM 100, EM 100 (salt), EM 112, EM 144, EM 612, Electol MM 2, HyQual, JPM 100, Kemilub EM-F-V, Liga MF 2V, M 5GN, Magnesium Stearate G, Magnesium distearate, Magnesium octadecanoate, Magnesium stearic acid, Mg-St, NP 1500, NS-M, NS-M (salt), Nissan Electol MM 2, PETS 120P, Palmstar 325BP/EP, Parteck LUB-MST, Petrac MG 20NF, Pharma, SM, SM 1000, SM-P, SPX 100F, Synpro 90, Synpro Magnesium Stearate 90
IUPAC Name magnesium;octadecanoate
SMILES [Mg+2].CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-]

Specifications

Quantity 500 g
InChI Formula InChI=1S/2C18H36O2.Mg/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2
Formula Weight 590.5125
Chemical Name or Material Magnesium Stearate (AS)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.