missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Hydrate, Purum p.a., ≥99.0% (calc. on dried material, KT), Honeywell™

Product Code. 15666080 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15666080 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15666080 Supplier Honeywell Research Chemicals Supplier No. 631391KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 1124°C
CAS 22189-08-8
Color Colorless
pH Neutral
Physical Form Solid
Quantity 1 kg
Molecular Formula MgO4S
Linear Formula MgSO4 · aq
MDL Number MFCD00149787
UN Number NONH for all modes of transport
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES [Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium;sulfate
Molecular Weight (g/mol) 120.36
PubChem CID 24083
ChEBI CHEBI:32599
Formula Weight 120.37g/mol (anhydrous basis)
Percent Purity ≥99.0% (calc. on dried material, KT)
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Magnesium sulfate hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.