Learn More
Macherey-Nagel™ MagnetaPure 32 Plus Automated DNA / RNA Purification System
Product Code. 18257428
Change view
Click to view available options
Quantity:
1 Piece
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18257428 1 Piece Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 18257428 Supplier Macherey-Nagel™ Supplier No. 747010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product is under eco-contribution tax.
MagnetaPure 32 Plus automated DNA / RNA purification system: Automated extraction and purification system for DNA and RNA, MN.
  • The MagnetaPure 32 Plus is an automated platform for magnetic bead-based DNA/RNA extraction with a minimal benchtop footprint
  • Capable of processing up to 32 samples simultaneously
  • The stand-alone instrument is easy to set up, program, and operate
  • Mixing, magnetic bead transfer, washing, and elution steps are performed automatically, saving valuable hands-on time
  • The purification process can be completed within 20 to 60 minutes depending on the type of NucleoMag kit used and the complexity of the program
Specifications
Volume (Metric) Processing 50 to 1000 μL
Capacity 1 to 32 Samples
Content And Storage 1 Piece
For Use With (Application) Automated extraction system for magnetic bead-based DNA/RNA purification
Quantity 1 Piece