Learn More
IKA™ Magnetic Stirrer
Product Code. 13450557 Shop All IKA Products
Change view
Click to view available options
Model:
RO 10
RO 15
RO 5
Stirring Positions:
5
10
15
Top Plate Surface Area (Metric):
120 x 470 mm
190 x 470 mm
280 x 470 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Stirring Positions Top Plate Surface Area (Metric) Unit Size
13450557 RO 10 10 190 x 470 mm Each
13430557 RO 15 15 280 x 470 mm Each
13440557 RO 5 5 120 x 470 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13450557 Supplier IKA™ Supplier No. 0003691000
  • ONLINE EXCLUSIVE PRICE
    1990.00€
    1863.00€ / Pack of 1
    Save 127.00€
    6% Off
View all promotions
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for synchronous stirring without heat. IKA Magnetic Stirrer includes magnetic coil technology to provide noiseless and consistent stirring on all positions. Speed remains constant, even when load changes.
Digital magnetic stirrer without heating, designed for synchronous stirring. Magnetic coil technology provides noiseless and consistent stirring on all positions. Speed remains constant, even when load changes. Closed and compact design allows easy cleaning and protects the equipment against the penetration of liquids. Additionally, a removable transparent anti-slip mat is included.
  • Adjustable reverse operation
  • Wear free magnetic coils
  • Error code display
  • Easy operation with touch keypad
  • Suitable for continuous operation
  • Speed range: 0 to 1200 rpm
TRUSTED_SUSTAINABILITY
Specifications
Speed 0 to 1200 rpm
Stirring Positions 10
Top Plate Material Stainless Steel
Voltage 100/240 V
Wattage 25 w
Height (Metric) 60 mm
Top Plate Surface Area (Metric) 190 x 470 mm
Width (English) 7.5 in.
Width (Metric) 190 mm
Amperage 1000 mA
Frequency 50/60 Hz
Model RO 10
Temperature Range (Metric) 5°C to 40°C
Type RO 10 Magnetic Stirrer
Weight (Metric) 5 kg