Learn More
MAL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-22522
Product Code. 18176125
  • 482.00€ / 100µg
Description
MAL Polyclonal specifically detects MAL in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAL
Polyclonal
Western Blot 1:500
mal, T-cell differentiation protein, myelin and lymphocyte protein, T-cell differentiation protein MAL, T-lymphocyte maturation-associated protein
Rabbit
Affinity Purified
RUO
4118
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS with 1 mg/ml BSA. with 0.05% Sodium Azide
MAL
Synthetic peptides corresponding to residues C(114)F D G F T Y (R/K) H Y H E N I(126) of human MAL.
100 μg
Primary
Human, Mouse, Rat, Xenopus species