Learn More
Rogo Sampaic™ Malassez Counting Cells
Product Code. 11720845
Change view
Click to view available options
Grid Type:
Double
Simple
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grid Type Unit Size
11720845 Double Each
11740835 Simple Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11720845 Supplier Rogo Sampaic™ Supplier No. ZMB021
  • ONLINE EXCLUSIVE PRICE
    64.85€
    51.05€ / Pack of 1
    Save 13.80€
    21% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For determining the number of particles per volume unit of a liquid. Rogo Sampaic™ Malassez Counting Cells have a simple or double grid with a depth of 0.2mm.
  • For blood samples
  • Provides for accurate manual measurements
Specifications
Product Type Malassez Cell
Grid Type Double
Quantity 1Each
Sample Type Blood
Depth (Metric) 0.2mm