Learn More
Hamilton™ Male Luer Lock / Threaded Connector (1/8 - NPT)
Description
Specification Sheet
- Connection 1 – Male Luer Lock
- Connection 2 – Male 1/8 NPT
- I.D. – 1.5 mm
Specifications
Specifications
| Type | Male Luer Lock |