Learn More
Value Plastics Male Luer Slip to 200 Series Barb
Product Code. 30054302 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/8 in.
3/32 in.
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Material (Seal) unitSize
30054302 1/16 in. Polypropylene Pack of 25
30053075 1/8 in. Polypropylene Pack of 25
30054143 3/32 in. Polypropylene Pack of 25
30054079 1/8 in. Nylon Pack of 25
30053226 1/16 in. Nylon Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30054302 Supplier Value Plastics Supplier No. 5012077
  • ONLINE EXCLUSIVE PRICE
    27.25€
    25.08€ / Pack of 25
    Save 2.17€
    7% Off
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
A secure connection under pressure with 200 series barbs
Quick and easy assembly and disassembly without the need for tools. Barbed ridges provide secure grip on the inside of flexible tubing. Slip luer fittings allow for a simple yet secure connection
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/16 in.
Inlet Male Luer Slip, Hose Barb
Material (Seal) Polypropylene