missing translation for 'onlineSavingsMsg'
Learn More

Maleimide, TRC

Product Code. 30473972
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
Unit Size:
10g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473972 2 g 2g
30482686 10 g 10g
30481827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473972 Supplier Toronto Research Chemicals Supplier No. TRCM1249802G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-59-3
Molecular Formula C4H3NO2
Molecular Weight (g/mol) 97.07
Synonym 2,5-Dioxo-2,5-dihydropyrrole, 3-Pyrroline-2,5-dione, Maleic Imide, NSC 13684
IUPAC Name pyrrole-2,5-dione
SMILES O=C1NC(=O)C=C1

Specifications

Quantity 2 g
InChI Formula InChI=1S/C4H3NO2/c6-3-1-2-4(7)5-3/h1-2H,(H,5,6,7)
Formula Weight 97.0164
Chemical Name or Material Maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.