missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malondialdehyde Antibody (11E3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59367F
This item is not returnable.
View return policy
Description
Malondialdehyde Monoclonal specifically detects Malondialdehyde in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Malondialdehyde | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μL | |
| Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins. | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 11000 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto