missing translation for 'onlineSavingsMsg'
Learn More

Maltose Monohydrate, Technical, SLR, Fisher Chemical™

Product Code. 10796291 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10796291 100 g 100g
10161620 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10796291 Supplier Fisher Chemical Supplier No. M/1450/48
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Maltose Monohydrate, Technical, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-79-4
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834,MFCD18804057
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym maltose
PubChem CID 134129532
IUPAC Name (2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-[(2R,3R,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Melting Point 130°C
Color White
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 360.29g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.