missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Antibody (3C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51671-0.025ml
This item is not returnable.
View return policy
Description
Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Mammaglobin A | |
| Monoclonal | |
| Unconjugated | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| Mouse | |
| 11 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| Ascites |
| Western Blot | |
| 3C8 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| SCGB2A2 | |
| Purified recombinant fragment of human SCGB2A2 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 4250 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction