Learn More
Mammaglobin A Antibody (CL8892) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05504UV
Product Code. 30295102
  • 737.00€ / 0.10mL
Description
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mammaglobin A
Monoclonal
DyLight 350
mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1
This antibody was developed using a synthetic peptide derived from Q13296, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL8892
50mM Sodium Borate
Mouse
Protein A purified
RUO
4250
Store at 4⪚C in the dark.