missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride, Tetrahydrate, MP Biomedicals

Product Code. 11477612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477612 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477612 Supplier MP Biomedicals Supplier No. 0215533480
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Crystalline form of manganese(II) chloride tetrahydrate used in various chemical applications.

Specifications

CAS 13446-34-9
Physical Form Crystalline
Quantity 100 g
Molecular Formula Cl2H8MnO4
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]
IUPAC Name manganese(2+) tetrahydrate dichloride
Molecular Weight (g/mol) 197.90
Formula Weight 167.27
Grade Research
Chemical Name or Material Manganese Chloride Tetrahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Manganese Chloride Tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Manganese Chloride Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.