missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Chloride Tetrahydrate, ≥99%, Reagent Grade, Honeywell™

Product Code. 15617950 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15617950 100 g Poly bottle 100g
15627950 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15617950 Supplier Honeywell Research Chemicals Supplier No. M3634100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Pink
pH 4.0 to 6.0 (at 20°C)
Physical Form Crystalline
Quantity 100 g
Linear Formula MnCl2 · 4H2O
UN Number NONH for all modes of transport
Solubility Information Completely soluble in water
Formula Weight 197.91g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly bottle
Density 1.913g/cu.cm
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.