Learn More
Mecmesin™ Manual Control Digital Control Tester
Product Code. 11923998
Encompass
Change view
Click to view available options
Manual Control Digital Control Tester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11923998 Manual Control Digital Control Tester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11923998 Supplier Mecmesin™ Supplier No. 876107
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Includes
Power adaptor, five AAA batteries
Accurate, low-cost digital torque tester.
  • Portable and easy to use
  • Low-cost digital torque tester
  • Fits any shaped sample upto 19.05cm
  • Low maintenance
  • Field test capabilities with protective splash-proof housing
  • Capacity: 60kg
  • Force:6N
  • Sampling rate: 5000 Hz
  • Power (VAC): 115
Specifications
Product Type Manual Control Digital Torque Tester
Description Force: 6N
For Use With (Application) Calculates torque