Learn More
Macherey-Nagel™ Manual Ergonomic Decapper
Product Code. 12808595 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
For 11mm Aluminum Crimp Caps
For 20mm Aluminum Crimp Caps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12808595 For 20mm Aluminum Crimp Caps Each
12898585 For 11mm Aluminum Crimp Caps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12808595 Supplier Macherey-Nagel™ Supplier No. 735320
  • ONLINE EXCLUSIVE PRICE
    274.23€
    267.75€ / Pack of 1
    Save 6.48€
    2% Off