missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558272
This item is not returnable.
View return policy
Description
Immunogen sequence: EAPLAKDGVL TLANNVTPAK DVPPLSETEA TPVPIKDMEI AQTQKGISED SHLESLQDVG QSAAPTFMIS PETVTGT Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 45%.
The protein encoded by this gene is a major non-neuronal microtubule-associated protein. This protein contains a domain similar to the microtubule-binding domains of neuronal microtubule-associated protein (MAP2) and microtubule-associated protein tau (MAPT/TAU). This protein promotes microtubule assembly, and has been shown to counteract destabilization of interphase microtubule catastrophe promotion. Cyclin B was found to interact with this protein, which targets cell division cycle 2 (CDC2) kinase to microtubules. The phosphorylation of this protein affects microtubule properties and cell cycle progression. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq].
Specifications
| MAP4 | |
| Polyclonal | |
| Unconjugated | |
| MAP4 | |
| AA407148; MAP 4; Map4; MAP-4; microtubule associated protein 4; microtubule-associated protein 4; Mtap4; Mtap-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4134 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P27816 | |
| MAP4 | |
| Recombinant protein corresponding to Human MAP4. Recombinant protein control fragment (Product #RP-96264). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction