missing translation for 'onlineSavingsMsg'
Learn More

Maprotiline EP Impurity C (as Hydrochloride), Mikromol™

Product Code. 30052588
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052588 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052588 Supplier Mikromol Supplier No. MM0820.030025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Maprotiline EP Impurity C (as Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine Hydrochloride
CAS 92202-51-2
Analyte or Component Names 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H21 N . Cl H
Certifications/Compliance ISO 9001
Synonym Maprotiline Hydrochloride Imp. C (EP), Maprotiline Imp. C (EP), 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine Hydrochloride, Maprotiline Hydrochloride Imp. C (EP) as Hydrochloride, Maprotiline Hydrochloride Impurity C as Hydrochloride, Maprotiline Impurity C as Hydrochloride
InChI Formula InChI=1S/C19H21N.ClH/c20-13-5-11-19-12-10-14(15-6-1-3-8-17(15)19)16-7-2-4-9-18(16)19;/h1-4,6-9,14H,5,10-13,20H2;1H
SMILES Cl.NCCCC12CCC(c3ccccc13)c4ccccc24
IUPAC Name 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine hydrochloride
Molecular Weight (g/mol) 299.84
Quantity 25 mg
Formula Weight 299.1441 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.