Learn More
Euro Protection™ Marble High-Cut Safety Shoes
Product Code. 12058426
Change view
Click to view available options
Size:
38
41
42
43
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12058426 43 Each
12008426 38 Each
12038426 41 Each
12048426 42 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12058426 Supplier Euro Protection™ Supplier No. 9MARH43
  • ONLINE EXCLUSIVE PRICE
    168.00€
    80.35€ / Pack of 1
    Save 87.65€
    52% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Euro Protection™ Marble High-Cut Safety Shoes have a padded collar and bellows tongue for a snug fit and include a composite toe cap for protection.
  • Padded collar and bellows tongue for snug fit.
  • Footbridge supplying torsional rigidity.
  • Nubuck leather upper.
  • Moisture wicking non-woven lining.
  • Removable, textile and foam insole.
  • Composite toe cap.
  • Non-metallic (composite fabric) puncture resistant sole.
  • Injected polyurethane midsole.
  • Polyurethane outsole.
  • A spare pair of shoelaces.
Specifications
Color Brown
Liner Material Moisture wicking non-woven lining.
Material Leather
Size 43
Sole Features Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole.
Sole Material Polyurethane, Composite Fabric
Toe Material Composite
Weight (Metric) 694 g per shoe in size 8
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Type Safety Shoe