missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARCH5 Recombinant Rabbit Monoclonal Antibody (1O4S1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557968
This item is not returnable.
View return policy
Description
Immunogen sequence: LNSIFPGIGC PVPRIPAEAN PLADHVSATR ILCGALVFPT IATIVGKLMF SSVNSNLQRT ILGGIAFVAI KGAFKVYFKQ QQYLRQAHRK ILNYPEQEEA.
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2.
Specifications
| MARCH5 | |
| Recombinant Monoclonal | |
| 0.58 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q3KNM2, Q9NX47 | |
| MARCH5 | |
| A synthetic peptide corresponding to a sequence within amino acids 179-278 of human MARCH5 (NP_060294.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1O4S1 | |
| Unconjugated | |
| MARCH5 | |
| 1810015H18Rik; 2310008I22Rik; 2700055A20Rik; 5730499H23Rik; E130202O05Rik; E3 ubiquitin-protein ligase MARCH5; March5; Marchv; MARCH-V; membrane associated ring finger 5; membrane associated ring-CH-type finger 5; membrane-associated ring finger (C3HC4) 5; membrane-associated RING finger protein 5; Membrane-associated RING-CH protein V; mitochondrial ubiquitin ligase; MITOL; RING finger protein 153; RING-type E3 ubiquitin transferase MARCH5; RNF153 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 294079, 54708, 69104 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction