missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00€ - 572.00€
Specifications
| Antigen | MARCH8 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18477931
|
Novus Biologicals
NBP1-85540-25ul |
25 μL |
292.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18298057
|
Novus Biologicals
NBP1-85540 |
0.1 mL |
572.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
44263 Polyclonal specifically detects 44263 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MARCH8 | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Cellular modulator of immune recognition, EC 6.3.2, EC 6.3.2.-, MARCH-VIIIcellular modulator of immune recognition, membrane-associated ring finger (C3HC4) 8, Membrane-associated RING finger protein 8, Membrane-associated RING-CH protein VIII, MIR, RING finger protein 178 | |
| MARCHF8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q5T0T0 | |
| 220972 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VQCKVYVQLWKRLKAYNRVIYVQNCPETSKKNIFEKSPLTEPNFENKHGHGICHSDTNSSCCTEPEDTGAEIIHV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 33 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title