missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Marcks Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10004-2-IG-20UL
This item is not returnable.
View return policy
Description
MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.Specifications
| Marcks | |
| Polyclonal | |
| Unconjugated | |
| Marcks | |
| Macs, Marcks, PKCSL | |
| Rabbit | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.95 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P26645 | |
| Marcks | |
| Protein A | |
| RUO | |
| 17118 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction