Learn More
MARK3 Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805790
Description
The protein encoded by this gene is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for this gene.Specifications
| MARK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P27448 | |
| MARK3 | |
| Human recombit protein fragment corresponding to amino acids 473-729 of human MARK3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D9 | |
| Unconjugated | |
| MARK3 | |
| CTAK1, KP78, PAR1A, Par-1a | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4140 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.