Learn More
MARK3 Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805790S
Description
MARK3 Monoclonal antibody specifically detects MARK3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MARK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NWZ8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54960 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D9 | |
| Unconjugated | |
| GEMIN8 | |
| 1600015G02Rik; A430080F22Rik; cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 2; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; ETK1; ETK-1; KP78; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 3; MARK3; microtubule affinity regulating kinase 3; Mpk10; MPK-10; PAR1A; Par-1a; Protein kinase STK10; ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78 | |
| Full length human recombinant protein of human GEMIN8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.