missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MASA Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99161G
This item is not returnable.
View return policy
Description
MASA Polyclonal antibody specifically detects MASA in Human samples. It is validated for Western Blot
Specifications
| MASA | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| acireductone synthase, DKFZp586M0524, E1, EC 3.1.3.77, enolase-phosphatase 1, enolase-phosphatase E1, MASA homolog, MASAFLJ12594, MST1452,3-diketo-5-methylthio-1-phosphopentane phosphatase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MASA | |
| 0.1 mL | |
| Stem Cell Markers | |
| 58478 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction