missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Maspin Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148576
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SERPINB5 is a tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.
Specifications
| Maspin | |
| Polyclonal | |
| Unconjugated | |
| SERPINB5 | |
| 1110036M19Rik; AI462524; AI646751; Maspin; ovalbumin; peptidase inhibitor 5; PI5; PI-5; protease inhibitor 5; protease inhibitor 5 (maspin); serine (or cysteine) peptidase inhibitor, clade B, member 5; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5; serine (or cysteine) proteinase inhibitor, clade B, member 5; serine protease inhibitor 7; serpin B5; serpin family B member 5; serpin peptidase inhibitor, clade B (ovalbumin), member 5; SERPINB5; Spi5; Spi7 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 5268 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P36952 | |
| SERPINB5 | |
| E. coli-derived recombinant human Serpin B5/Maspin, Met1-Pro375. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction