Learn More
Atago™ MASTER-20M Refractometer
Product Code. 11416148
Change view
Click to view available options
Scale Range:
0 to 20 Brix
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scale Range Unit Size
11416148 0 to 20 Brix Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11416148 Supplier Atago™ Supplier No. 2383
  • ONLINE EXCLUSIVE PRICE
    128.00€
    99.00€ / Pack of 1
    Save 29.00€
    22% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Easy-to-read, easy-to-hold, and smooth-sampling refractometer. Atago™ MASTER-20M Refractometer is a low-concentration model.
  • Low concentration
  • Easy disposal of sample from the edge
  • Textured grip
  • Well balanced
  • Streamlined body
  • Easily cleaned with running water
  • Convenient storage case
  • Automatic sample distribution
  • Sample size: 2-3 drops
Specifications
Type Refractometer
Scale Range 0 to 20 Brix
Model MASTER-20M
Accuracy ±0.2% Brix
Repeatability ±0.1% Brix
Scale Type Brix
For Use With (Application) Measurements of low concentration fruit juices, cutting oil
Weight (Metric) 165 g
Dimensions (L x W x H) 20.7 x 3.2 x 3.4 cm