Learn More
Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate
Product Code. 13475339
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
No. per Pack:
1
Bulk
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility No. per Pack unitSize
13475339 Non-sterile Bulk Pack of 50
10013381 Sterile 1 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13475339 Supplier Greiner Bio-One Supplier No. 780210
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate is also suitable for storing sample material.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films and heat sealer
  • High temperature resistance
  • High chemical resistance
  • A
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 1 mL
Well Shape Round Bottom (U-well)
No. per Pack Bulk
Quantity 50/Cs.
Well Design Chimney