Learn More
Masterflex™ Tygon™ E-Lab (E-3603) Pump Tubing
Product Code. 13528990 Shop All Masterflex Products
Change view
Click to view available options
Tubing Size:
L/S 13
L/S 14
L/S 15
L/S 16
L/S 17
L/S 18
L/S 25
L/S 36
L/S 73
Unit Size:
15m
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
13528990 L/S 13 Each
13558990 L/S 14 Each
13585610 L/S 15 15m
13538990 L/S 16 15m
13568990 L/S 17 15m
13518990 L/S 18 15m
13548990 L/S 25 15m
13598990 L/S 36 Each
13529000 L/S 73 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 13528990 Supplier Masterflex™ Supplier No. 0650913
  • ONLINE EXCLUSIVE PRICE
    85.00€
    69.00€ / Pack of 1
    Save 16.00€
    18% Off
View all promotions
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use this tubing for general laboratory applications. Tygon™ E-Lab (E-3603) Pump Tubing is ideal for inorganic chemicals and viscous fluids.
  • Clear tubing for easy flow monitoring
  • Nonaging, nonoxidizing
  • Low gas permeability
  • High dielectric constant
Specifications
Diameter (Metric) Inner 0.8 mm
Material Tygon
Overall Length (Metric) 1500 cm
For Use With (Application) For general transfer applications
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 1 Each
Tubing Size L/S 13
Flow Rate 0.06 to 36mL/min.
Hose Barb 1/16in.
Height (Metric) 8.8 m
Color Clear
Maximum Pressure 40 psi
Vacuum Level 140 mbar
Max. Operating Temperature (Metric) 74°C
Product Type Pump Tubing