missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MASTL Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97307H
This item is not returnable.
View return policy
Description
MASTL Polyclonal antibody specifically detects MASTL in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MASTL | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| HRP | |
| FLJ14813EC 2.7.11.1, GREATWALL, GW, GWL, hGWL, MAST-L, microtubule associated serine/threonine kinase-like, Microtubule-associated serine/threonine-protein kinase-like, RP11-85G18.2, THC2greatwall protein kinase | |
| Produced in rabbits immunized with E. coli-derived Human MASTL fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 84930 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction