missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT1A Antibody (4D11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00004143-M01
This item is not returnable.
View return policy
Description
MAT1A Monoclonal antibody specifically detects MAT1A in Human, C. elegans samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| MAT1A | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| AdoMet synthase 1, adoMet synthetase 1, AMS1, MAT, MATA1MAT 1, MAT-I/III, Methionine adenosyltransferase 1, methionine adenosyltransferase I, alpha, Methionine adenosyltransferase I/III, S-adenosylmethionine synthase isoform type-1, S-adenosylmethionine synthetase isoform type-1, SAMS, SAMS1EC 2.5.1.6 | |
| MAT1A (AAH18359, 1 a.a. ~ 395 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNGPVDGLCDHSLSEGVFMFTSESVGEGHPDKICDQISDAVLDAHLKQDPNAKVACETVCKTGMVLLCGEITSMAMVDYQRVVRDTIKHIGYDDSAKGFDFKTCNVLVALEQQSPDIAQCVHLDRNEEDVGAGDQGLMFGYATDETEECMPLTIILAHKLNARMADLRRSGLLPWLRPDSKTQVTVQYMQDNGAVIPVRIHTIVISVQHNEDITLEEMRRALKEQVIRAVVPAKYLDEDTVYHLQPSGRFVIGGPQGDAGVTGRKIIVDTYGGWGAHGGGAFSGKDYTKVDRSAAYAARWVAKSLVKAGLCRRVLVQVSYAIGVAEPLSISIFTYGTSQKTERELLDVVHKNFDLRPGVIVRDLDLKKPIYQKTACYGHFGRSEFPWEVPRKLVF | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 4143 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunoprecipitation | |
| 4D11 | |
| Western Blot 1:500, ELISA, Immunoprecipitation 1:10-1:500 | |
| AAH18359 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, C. elegans | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction