missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATN3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77022
This item is not returnable.
View return policy
Description
MATN3 Polyclonal specifically detects MATN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| MATN3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DIPOA, EDM5, HOA, matrilin 3, matrilin-3, OADIP, OS2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4148 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2.5-20 μg/mL | |
| AAI39908 | |
| MATN3 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the center of human MATN3. The immunogen is located within amino acids 180 - 230 of MATN3. Amino Acid Squence: VTDGRPQDQVNEV | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction