missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD1 Antibody (CPTC-MBD1-1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08387AF750
This item is not returnable.
View return policy
Description
MBD1 Monoclonal antibody specifically detects MBD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MBD1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription | |
| Recombinant human MBD1 protein (Uniprot: Q9UIS9) | |
| 100 μg | |
| Chromatin Research | |
| 4152 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CPTC-MBD1-1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction