Learn More
Invitrogen™ MBD1 Recombinant Rabbit Monoclonal Antibody (24GB405)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557470
 View more versions of this product
Product Code. 30274805
  • 564.00€ / 100µL
Description
MBD1 Recombinant Monoclonal Antibody for Western Blot, Flow
Methyl-CpG-binding domain protein 1 (MBD1) is a member of a family of mammalian proteins that specifically bind methylated regions of DNA. It is a transcriptional repressor that binds CpG islands in promoters where the DNA is methylated at position 5 of cytosine within CpG dinucleotides. Binding is abolished by the presence of 7-mG that is produced by DNA damage by methylmethanesulfonate (MMS). MBD1 acts as a transcriptional repressor and plays a role in gene silencing by recruiting AFT7IP, which in turn recruits factors such as the histone methyltransferase SETDB1. Isoform 1 and isoform 2 can also repress transcription from unmethylated promoters.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MBD1
Recombinant Monoclonal
Unconjugated
MBD1
CXXC3; CXXC-type zinc finger protein 3; Mbd1; methyl-CpG binding domain protein 1; methyl-CpG binding domain protein 1 isoform PCM1; methyl-CpG-binding domain protein 1; Methyl-CpG-binding protein MBD1; PCM1; Protein containing methyl-CpG-binding domain 1; RFT; the regulator of fibroblast growth factor 2 (FGF-2) transcription
Rabbit
Affinity chromatography
RUO
291439, 4152
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB405
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UIS9
MBD1
A synthesized peptide derived from human MBD1 (aa 550-605).
100 μL
Primary
Human, Rat