missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBL Antibody (11F11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05078
This item is not returnable.
View return policy
Description
MBL Monoclonal antibody specifically detects MBL in Human, Rat samples. It is validated for Western Blot, ELISA
Specifications
| MBL | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:2000 | |
| COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBLmannan-binding lectin, MBP, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833 | |
| MBL purified from human donor plasma and adsorbed onto aluminum hydroxide. | |
| 0.1 mg | |
| Immunology | |
| 4153 | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 11F11 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction