missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (CL2829) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46632AF405
This item is not returnable.
View return policy
Description
MBP Monoclonal antibody specifically detects MBP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MBP | |
| Monoclonal | |
| Alexa Fluor 405 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| This antibody was developed using a recombinant protein derived from P02686, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4155 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2829 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction