Learn More
Gibco™ MBP Bovine Protein, Natural
Product Code. 11578866 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11578866 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11578866 Supplier Gibco™ Supplier No. 13228010
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MBP Bovine Protein, Natural
Myelin Basic Protein (MBP) was purified from bovine brain by a modification of the method of Deibler et al. (1). The protein is supplied lyophilized.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Species Bovine
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Approved for shipment on Wet or Dry Ice
Form Powder