missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBTPS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00€ - 572.00€
Specifications
| Antigen | MBTPS2 |
|---|---|
| Concentration | 0.05mg/mL |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18427420
|
Novus Biologicals
NBP1-80800-25ul |
25ul |
292.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18289155
|
Novus Biologicals
NBP1-80800 |
0.1 mL |
572.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBTPS2 Polyclonal specifically detects MBTPS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MBTPS2 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O43462 | |
| 51360 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EVAEDSPAIGPRGLFVGDLVTHLQDCPVTNVQDWNECLDTIAYEPQIGYCISASTLQQLSFPVRAYKRLDGSTECCNNHSLTDVCFSYRNNFNKRLHTCLPARKAVEATQVCRTNKDCKKSSSSSFCIIPSLETHTR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.05mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 3.4.24.85, Endopeptidase S2P, FLJ32174, membrane-bound transcription factor peptidase, site 2, membrane-bound transcription factor protease, site 2, membrane-bound transcription factor site-2 protease, S2PIFAP, SREBPs intramembrane protease, Sterol regulatory element-binding proteins intramembrane protease | |
| MBTPS2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title