missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (CL12855), Novus Biologicals™
Mouse Monoclonal Antibody
329.00€ - 649.00€
Specifications
| Antigen | MCAM/CD146 |
|---|---|
| Clone | CL12855 |
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650415
|
Novus Biologicals
NBP3-18546-100ul |
100 μg |
649.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677324
|
Novus Biologicals
NBP3-18546-25ul |
25 μg |
329.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MCAM/CD146 | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
| This antibody was developed against a recombinant protein of P43121. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12855 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 4162 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title