missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563542
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MMDSEAHEKR PPILTSSKQD ISPHITNVGE MKHYLC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 69%.
SLC25A51 gene ontology annotations related to this gene include transmembrane transporter activity.Specifications
MCAR1 | |
Polyclonal | |
Unconjugated | |
SLC25A51 | |
9130208E07Rik; CG7943; D130005A03Rik; Gm138; Mcart1; mitochondrial carrier triple repeat 1; mitochondrial carrier triple repeat protein 1; mitochondrial carrier triple repeat protein 1-like; SLC25A51; Solute carrier family 25 member 51; solute carrier family 25, member 51 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92014 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1U9 | |
SLC25A51 | |
Recombinant protein corresponding to Human MCAR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only