missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mcl-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56146
This item is not returnable.
View return policy
Description
Mcl-1 Polyclonal antibody specifically detects Mcl-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Mcl-1 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| bcl2-L-3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM | |
| A synthetic peptide corresponding to amino acids 121-139 (SPEEELDGYEPEPLGKRPA) of human Mcl-1 was used as immunogen, GenBank no. NP_068779.1. The immunogen sequence is 100% conserved in Mcl-1 isoform 1 (GenBank no. NP_068779.1) and isoform 2 (GenBank no. NP_877495.1). | |
| 0.05 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 4170 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction