Learn More
Lovibond™ MD200 4-in-1 photometer kit for chlorine pH cyanuirc acid and M-alkalinity with tablet reagents
Product Code. 12671725
Change view
Click to view available options
Product Type:
Photometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12671725 Photometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 12671725 Supplier Lovibond™ Supplier No. 2860502
  • ONLINE EXCLUSIVE PRICE
    750.00€
    661.02€ / Pack of 1
    Save 88.98€
    11% Off
Estimated Shipment: 28-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
includes tablet reagents, 3 round vials with lids, carry case, batteries and instruction manual
Versatile, compact and: The benchtop photometer MD 200 combines high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic. The result: precise measurement results at all times - no matter where you use it.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Product Type Photometer
Display Type LCD
For Use With (Application) Chlorine, pH, Cyanuirc Acid and M-Alkalinity Measurement