Learn More
MDC1 Antibody (P2B11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12890
This item is not returnable.
View return policy
Description
MDC1 Monoclonal specifically detects MDC1 in Human, Mouse, Bovine, Non-human Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MDC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| MDC1 | |
| GST-tagged recombinant protein corresponding to mouse MDC1 at and around the N-terminus | |
| 0.1 mg | |
| Primary | |
| Detects approx 184kDa. This antibody recognizes MDC1 at and around the N-terminus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| P2B11 | |
| Unconjugated | |
| DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 9656 | |
| Mouse | |
| Purified |