missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15462-1-AP-20UL
This item is not returnable.
View return policy
Description
MDH2 is localized to the mitochondria and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.Specifications
| MDH2 | |
| Polyclonal | |
| Unconjugated | |
| MDH2 | |
| M MDH, MDH, MDH2, MOR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17448, 4191, 81829 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04636, P08249, P40926 | |
| Mdh2 | |
| MDH2 Fusion Protein Ag7750 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction