missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2/HDM2 Antibody (MDM2/7189R), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21097IR
This item is not returnable.
View return policy
Description
MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MDM2/HDM2 | |
| Monoclonal | |
| DyLight 755 | |
| ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
| Recombinant human MDM2/HDM2 protein fragment (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors | |
| 4193 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MDM2/7189R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction