Learn More
MDM4 Monoclonal Antibody (OTI4G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505726
Description
Inhibits p53/TP53- and TP73/p73-mediated cell cycle arrest and apoptosis by binding its transcriptional activation domain. Inhibits degradation of MDM2. Can reverse MDM2-targeted degradation of TP53 while maintaining suppression of TP53 transactivation and apoptotic functions.Specifications
| MDM4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15151 | |
| MDM4 | |
| Full length human recombit protein of human MDM4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G3 | |
| Unconjugated | |
| MDM4 | |
| HDMX, MDMX, MRP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4194 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.