Learn More
MDM4 Monoclonal Antibody (OTI4G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505726S
Description
MDM4 Monoclonal antibody specifically detects MDM4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MDM4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15151 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4194 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G3 | |
| Unconjugated | |
| MDM4 | |
| 4933417N07Rik; AA414968; AL023055; AU018793; AU021806; C85810; DKFZp781B1423; double minute 4 homolog; double minute 4 protein; double minute 4, human homolog of; p53-binding protein; HDMX; human homolog of; Mdm2-like p53-binding protein; MDM4; Mdm4 p53 binding protein homolog; MDM4 protein variant G; MDM4 protein variant Y; MDM4, p53 regulator; MDM4-related protein 1; Mdmx; MGC132766; MRP1; p53-binding protein Mdm4; Protein Mdm4; protein Mdmx; RP11-430C7.1; transformed mouse 3T3 cell double minute 4 | |
| Full length human recombinant protein of human MDM4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.