missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDMX Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-556H
This item is not returnable.
View return policy
Description
MDMX Polyclonal antibody specifically detects MDMX in Human samples. It is validated for Western Blot
Specifications
| MDMX | |
| Polyclonal | |
| Western Blot | |
| DKFZp781B1423, double minute 4 homolog, Double minute 4 protein, double minute 4, human homolog of; p53-binding protein, HDMX, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein (mouse), MDM4-related protein 1, MDMXMGC132766, mouse double minute 4, human homolog of; p53-binding protein, MRP1, p53-binding protein Mdm4, protein Mdm4, Protein Mdmx | |
| The immunogen recognized by this antibody maps to a region between residues 125 and 175 of human ortholog of mouse double minute 4 using the numbering given in Swiss-Prot entry O15151 (GeneID 4194). | |
| 0.1 mL | |
| Apoptosis, DNA Repair, Phospho Specific, Signal Transduction | |
| 4194 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto