missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ME0328

Product Code. 17159991
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17159991 1 mg 1mg
17169991 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17159991 Supplier STEMCELL Technologies Supplier No. 1000271
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PARP-3 inhibitor

  • ME0328 is a cell-permeable and selective inhibitor of poly(ADP-ribose) polymerase 3 (PARP-3; IC50 = 0.89 μM; Lindgren et al.) PARP-3 is required for stabilization of the mitotic spindle and crucial for mitotic progression (Sharif-Askari et al.)
  • Enhances vinorelbine-induced microtubule destabilization, leading to cell cycle arrest and apoptosis in breast cancer cells (Sharif-Askari et al.)

Specifications

CAS 1445251-22-8
Quantity 1 mg
Molecular Formula C19H19N3O2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.